Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

L9669

Sigma-Aldrich

Anti-Livin (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-BIRC7, Anti-Baculoviral IAP repeat-containing 7, Anti-KIAP, Anti-MLIAP, Anti-RNF50

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~39 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using lysate of HEK-293T cells expressing human Livin
western blot: 2.5-5.0 μg/mL using whole extract of HEK-293T cells expressing human Livin

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BIRC7(79444)

General description

Livin or baculoviral IAP repeat containing 7 (BIRC7) gene is mapped to human chromosome 20q13.33 and is expressed in α and β isoforms. It belongs to the family of inhibitor of apoptosis proteins (IAP). Livin contains a single copy of a baculovirus IAP repeat (BIR) as well as a really interesting new gene (RING)-type zinc finger domain. Structurally, livin comprises a three-stranded anti-parallel β-sheet and four α-helices and takes a globular architecture.

Specificity

Anti-Livin (N-terminal) recognizes human Livin/BIRC7.

Application

Anti-Livin (N-terminal) antibody produced in rabbit may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Livin or baculoviral IAP repeat containing 7 (BIRC7) inhibitory functionality is mediated by the baculovirus inhibitor of apoptosis proteins (IAP) repeat (BIR) domain. This domain is also essential for interaction with caspases. The really interesting new gene (RING) finger domain elicits E3-ubiquitin ligase activity in the absence of BIR domain. The two isoforms of livin have different antiapoptotic properties, with isoform α; protecting cells from apoptosis induced by staurosporine and isoform β protecting cells from apoptosis induced by etoposide. An elevated expression of Livin is detected in prostate, breast, colon, head and neck squamous cell carcinoma. Inhibition of livin in nasopharyngeal carcinoma (NPC) leads to improved irradiation sensitivity in NPC cells.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boaz Nachmias et al.
Cancer research, 63(19), 6340-6349 (2003-10-16)
Inhibitor of apoptosis protein (IAP) is a family of intracellular proteins that plays an essential role in the regulation of apoptosis. Recently, we and others discovered a new member of this family, termed Livin. Many studies have focused on the
Tae Mi Yoon et al.
Oncology reports, 37(6), 3667-3673 (2017-04-26)
The responsiveness of head and neck squamous cell carcinoma (HNSCC) to chemotherapy widely affects prognosis. Overcoming chemoresistance is necessary to improve prognoses in patients with advanced HNSCC. Evasion of apoptosis by cancer cells is a major cause of chemoresistance. Livin
Fei Ma et al.
PloS one, 15(3), e0229272-e0229272 (2020-03-03)
Radiotherapy is one of the major remedies for the treatment of cancer, including nasopharyngeal carcinoma (NPC). Radioresistance occurs very often in target cells that is a large drawback in cancer treated with radiotherapy. Livin involves the over-growth of cancer cells.
Irena Crnković-Mertens et al.
Journal of molecular medicine (Berlin, Germany), 84(3), 232-240 (2006-01-27)
Livin (alternatively called ML-IAP or KIAP) is a cancer-associated member of the antiapoptotic inhibitor of apoptosis protein family. Two splicing variants of Livin, designated Livin alpha and Livin beta, have been identified. The significance of these isoforms for Livin-mediated apoptosis
Wei-Na Zhang et al.
Oncotarget, 7(52), 87449-87461 (2016-12-16)
Even though infection with human papillomaviruses (HPV) is very important, it is not the sole cause of cervical cancer. Because it is known that genetic variations that result from HPV infection are probably the most important causes of cervical cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service