Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L9514

Sigma-Aldrich

Ricin A chain from Ricinus communis (castor bean)

buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

form

buffered aqueous glycerol solution

Quality Level

potency

≥250 μg/mL per mL agglutination activity

composition

Protein, ≥0.3 mg/mL Lowry-TCA

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ricin A chain inactivates the 60 S ribosomal subunit of eukaryotic cells, inhibiting protein synthesis. This toxic activity is independent of the presence of the B chain of ricin.

Other Notes

Ricinus communis toxin (also named RCA60, RCAII, Ricin D, or RCL III) is a highly toxic protein, mol. wt. ~60 kDa, composed of two chains, A and B, connected by a single disulfide bridge.

Quality

Electrophoretically pure (SDS-PAGE)

Physical form

Solution in 40% glycerol containing 10 mM phosphate, pH 6.0, 0.15 M NaCl, 10 mM galactose and 0.5 mM dithioerythritol

Preparation Note

Purified by affinity chromatography.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 7.2, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hour incubation at 25°C.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Skin Sens. 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natália L de Sousa et al.
Journal, genetic engineering & biotechnology, 20(1), 16-16 (2022-01-29)
Castor bean (Ricinus communis L.) is an important cultivated oilseed. Seeds contain ricinoleic acid, a valuable product for a variety of industries. Castor cake is a residue of ricinoleic manufacture and could be used as animal feed due to its
Lisa Fetter et al.
Chemical communications (Cambridge, England), 51(82), 15137-15140 (2015-09-02)
Protein toxins present considerable health risks, but detection often requires laborious analysis. Here, we developed electrochemical aptamer biosensors for ricin and botulinum neurotoxins, which display robust and specific signal at nanomolar concentrations and function in dilute serum. These biosensors may
Staci Kane et al.
Journal of immunological methods, 451, 54-60 (2017-09-01)
With several ricin contamination incidents reported over the past decade, rapid and accurate methods are needed for environmental sample analysis, especially after decontamination. A sample processing method was developed for common surface sampling devices to improve the limit of detection
Ross Thyer et al.
Nature biotechnology, 36(7), 624-631 (2018-06-05)
Incorporation of the rare amino acid selenocysteine to form diselenide bonds can improve stability and function of synthetic peptide therapeutics. However, application of this approach to recombinant proteins has been hampered by heterogeneous incorporation, low selenoprotein yields, and poor fitness
Natália L Sousa et al.
Scientific reports, 7(1), 15385-15385 (2017-11-15)
Ricin is a highly toxic ribosome-inactivating lectin occurring in the seeds of castor bean (Ricinus communis L.). Castor bean grows throughout tropical and sub-tropical regions and is a very important crop due to its high seed content of ricinoleic acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service