Recommended Products
application(s)
environmental
food and beverages
microbiology
shipped in
wet ice
storage temp.
2-8°C
suitability
Legionella spp.
Application
An enrichment supplement recommended for enhanced growth of Legionella species.
Other Notes
Composition: (per vial)
L-Cysteine hydrochloride: 200.00 mg
Ferric pyrophosphate: 125.00 mg
L-Cysteine hydrochloride: 200.00 mg
Ferric pyrophosphate: 125.00 mg
Reconstitution
Sufficient for 500 ml medium
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service