Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L8413

Sigma-Aldrich

Leucozyme from human sputum

enzyme mixture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

solid

solubility

soluble

storage temp.

2-8°C

General description

Leucozyme is a spheroplasting agent. [1]
at pH 3-9 in low molarity buffers (<0.05 M NaCl). Solubility increases with increased NaCl concentration.

Application

Leucozyme has been used in a study to investigate defense mechanisms against infection. [2]

Physical form

Contains elastase (approx 5%), myeloperoxidase (6%), lysozyme (1%), trace cathespin G, trypsin and other proteins.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

On the Defence Mechanisms Against Infection
Amano, T.
Defense Technical Information Center (1962)
Nature of spheroplasting agent, leucozyme C, in guinea pig leucocytes.
M Ozaki et al.
Biken journal, 8(4), 175-188 (1965-12-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service