Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L8141

Sigma-Aldrich

Anti-Human Lambda Light Chain (Bound and Free) antibody produced in rabbit

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

secondary antibodies

clone

polyclonal

contains

15 mM sodium azide

technique(s)

quantitative precipitin assay: 1.5

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Lambda (λ) light chain has an amino-terminal variable region and carboxy-terminal constant region. λ chain is biologically important and constitutes 30 to 40% of human immunoglobulin (Ig) molecules. The human λ locus is mapped to human chromosome 22.

Biochem/physiol Actions

Immunoglobulins (Ig) are produced by the B cells and play crucial role in host humoral immune response. A typical immunoglobulin consists of two heavy chains and two light chains. The two light chains are either κ or the λ chain. The B cells express one type of light chain during their life time.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B-cell development
Hematology (2018)
Structural and functional properties of human lambda-light-chain variable-region subgroups.
A Solomon et al.
Clinical and diagnostic laboratory immunology, 2(4), 387-394 (1995-07-01)
Jerry A Katzmann et al.
Clinical chemistry, 48(9), 1437-1444 (2002-08-27)
The detection of monoclonal free light chains (FLCs) is an important diagnostic aid for a variety of monoclonal gammopathies and is especially important in light-chain diseases, such as light-chain myeloma, primary systemic amyloidosis, and light-chain-deposition disease. These diseases are more

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service