Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L8025

Sigma-Aldrich

Lectin from Maackia amurensis

lyophilized powder

Synonym(s):

Maackia amurensis agglutinin, MAA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

form

lyophilized powder

potency

<10 μg per mL agglutination activity

composition

Protein, ≥85% Lowry

impurities

salt, essentially free

storage temp.

2-8°C

Biochem/physiol Actions

MAA consists of two molecular species, a strongly hemagglutinating hemagglutinin (MAH) and a strongly mitogenic hemagglutinin (MAL). The mitogenic activity of the latter is inhibited by α-sialyl-[2→3]-lactose. MAA interacts with sialic acid-containing glycoconjugate and may be useful for fractionation of sialylated oligosaccharides.

Preparation Note

Purified by affinity chromatography using a modification of the method of Wang and Cummings.[1]

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 7.2 of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group O erythrocytes after 1 hr incubation at 25 °C.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W C Wang et al.
The Journal of biological chemistry, 263(10), 4576-4585 (1988-04-05)
We recently reported that the purified leukoagglutinin (designated MAL) from the seeds of the leguminous plant Maackia amurensis is a potent leukoagglutinin for the mouse lymphoma cell line BW5147 (Wang, W.-C., and Cummings, R. D. (1987) Anal. Biochem. 161,80). We
Jhon Alberto Ochoa-Alvarez et al.
PloS one, 7(7), e41845-e41845 (2012-07-31)
Cancer is a leading cause of death of men and women worldwide. Tumor cell motility contributes to metastatic invasion that causes the vast majority of cancer deaths. Extracellular receptors modified by α2,3-sialic acids that promote this motility can serve as

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service