Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L6508

Sigma-Aldrich

L-Lactic dehydrogenase LDH-5 (M4) Isoenzyme from Human Placenta

Type XXVIII, ammonium sulfate suspension, 300-600 units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

biological source

human placenta

type

Type XXVIII

form

ammonium sulfate suspension

specific activity

300-600 units/mg protein

foreign activity

pyruvate kinase, essentially free

Storage temp.

2-8°C

Unit Definition

One unit will reduce 1.0μmole of pyruvate to L-lactate per min at pH7.5 at 37 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4 solution, pH 6.0

Analysis Note

Protein determined by biuret.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service