L6043
Monoclonal Anti-LAP2 (TMPO) antibody produced in mouse
~1 mg/mL, clone LAP106, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-CMD1T, Anti-Cardiomyopathy, dilated, 1T, included, Anti-Lamina Associated Polypeptide 2, Anti-TMPO, Anti-Thymopoietin
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
LAP106, monoclonal
form
buffered aqueous solution
mol wt
~50 kDa
species reactivity
human, mouse, canine
concentration
~1 mg/mL
technique(s)
immunocytochemistry: suitable
indirect ELISA: suitable
western blot: 2-4 μg/mL using HeLa nuclear cell extract
isotype
IgG1
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... TMPO(7112)
mouse ... Tmpo(21917)
Specificity
Immunogen
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
related product
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service