Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L5640

Sigma-Aldrich

Lectin from Agaricus bisporus (mushroom)

lyophilized powder

Synonym(s):

Agaricus bisporus agglutinin, ABA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200

form

lyophilized powder

potency

<16 μg protein per mL agglutination activity

composition

Protein, ~10% biuret

storage temp.

−20°C

Biochem/physiol Actions

ABA is a mixture of two phytohemagglutinins with similar specificities for carbohydrate receptors. The lectin is not blood group specific, but has an affinity for fetuin.

Physical form

Lyophilized powder containing NaCl

Preparation Note

Purified by affinity chromatography.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group O erythrocytes after 1 hr incubation at 25 °C.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service