Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L5506

Sigma-Aldrich

L-Leucine−Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.56

form

saline suspension

extent of labeling

2-10 μmol per mL

matrix

4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

storage temp.

2-8°C

Application

L-leucine-agarose is used in protein chromatography, affinity chromatography and amino acid resins. L-leucine-agarose has been used to study pulp regeneration following pulp exposure as well as to compare polymeric and monomeric affinity ligands′ adsorption affinity, capacity and selectivity to oligopeptide.

Physical form

Suspension in 2.0 M NaCl

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Feng et al.
Journal of colloid and interface science, 355(2), 478-485 (2011-01-15)
This work aimed to compare two types of affinity ligands, i.e. polymeric and monomeric ligands, by investigating their adsorption affinity, capacity and selectivity to oligopeptide. The peptide NH(2)-VVRGCTWW-COOH (VW-8) was chosen as the target adsorbate, while histidine (His), aspartic acid
Amelogenin Exons 8 and 9 Encoded Peptide Enhances Leucine Rich Amelogenin Peptide Mediated Dental Pulp Repair
Huang, Y., et al.
Cells Tissues Organs (2012)
Marina Marcuschi et al.
Biochemical and biophysical research communications, 396(3), 667-673 (2010-05-05)
An enzyme was purified from the pyloric caecum of tambaqui (Colossoma macropomum) through heat treatment, ammonium sulfate fractionation, Sephadex G-75 and p-aminobenzamidine-agarose affinity chromatography. The enzyme had a molecular mass of 23.9 kDa, NH(2)-terminal amino acid sequence of IVGGYECKAHSQPHVSLNI and
Dong Hwan Ho et al.
Cell biochemistry and function, 36(8), 431-442 (2018-11-10)
Leucine-rich repeat kinase 2 (LRRK2) is a causal gene of Parkinson disease. G2019S pathogenic mutation increases its kinase activity. LRRK2 regulates various phenotypes including autophagy, neurite outgrowth, and vesicle trafficking. Leucyl-tRNA synthetase (LRS) attaches leucine to tRNALeu and activates mTORC1.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service