Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L5045

Sigma-Aldrich

Luotonin B

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11N3O2
CAS Number:
Molecular Weight:
301.30
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

synthetic (organic)

assay

≥98%

form

solid

solubility

chloroform/methanol (4:1): 2 mg/mL, clear to slightly hazy, colorless to yellow

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TOP1(7150)

General description

Luotonin B is a heteroaromatic pyrroloquinazolinoquinoline alkaloid that, along with Luotonin A and E, was originally isolated from the Chinese medicinal plant Paganum nigellastrum.[1] It is an oxidation product of Luotonin A and is converted to Luotonin E by methanol treatment.[1]

Biochem/physiol Actions

Luotonin compounds have antitumor activity and demonstrate cytotoxicity and inhibitory activity against human topoisomerase. For use as a topoisomerase inhibitor[1], a starting concentration of 25 μM is suggested. It is suggested that a dose-response curve be performed to find the inhibitor concentration most suitable for the intended assay.

Reconstitution

Solubility is tested at 2 mg/mL in 4:1 solution of choloform and methanol. Solubility of luotonins in DMSO has been reported.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances in the studies on luotonins
Liang JL, et al.
Molecules (Basel), 16(6), 4861-4883 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service