Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L4886

Sigma-Aldrich

Leukotriene C4

≥92.5% (HPLC), 45-70 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H47N3O9S
CAS Number:
Molecular Weight:
625.77
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥92.5% (HPLC)

concentration

45-70 μg/mL in methanol

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC(CCCC(O)=O)C(SCC(NC(CCC(C(O)=O)N)=O)C(NCC(O)=O)=O)/C=C/C=C/C=C\C/C=C\CCCCC

InChI

1S/C30H47N3O9S/c1-2-3-4-5-6-7-8-9-10-11-12-13-16-25(24(34)15-14-17-27(36)37)43-21-23(29(40)32-20-28(38)39)33-26(35)19-18-22(31)30(41)42/h6-7,9-13,16,22-25,34H,2-5,8,14-15,17-21,31H2,1H3,(H,32,40)(H,33,35)(H,36,37)(H,38,39)(H,41,42)/b7-6+,10-9+,12-11+,16-13+

InChI key

GWNVDXQDILPJIG-KVUXNHQJSA-N

Biochem/physiol Actions

Leukotriene formed from the conjugation of LTA4 with glutathione. Induces bronchoconstriction and renal vasoconstriction.

Packaging

Packaged under argon.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rikiya Koketsu et al.
International archives of allergy and immunology, 161 Suppl 2, 23-31 (2013-06-05)
Basophils and mast cells are important initiator/effector cells capable of rapidly responding to IgE-mediated stimulation, but the precise mechanisms regulating their functions in vivo have not been fully identified. In this study, we assessed whether low levels of antigen can
Gabriele Jedlitschky et al.
Vitamins and hormones, 64, 153-184 (2002-03-20)
Transport proteins control the release of the endogenous glutathione conjugate leukotriene C4 (LTC4) from leukotriene-synthesizing cells as well as its hepatobiliary and renal elimination. The photolabile conjugated triene structure of LTC4 has enabled direct photoaffinity labeling of the multidrug resistance
B Samuelsson
Science (New York, N.Y.), 220(4597), 568-575 (1983-05-06)
Arachidonic acid plays a central role in a biological control system where such oxygenated derivatives as prostaglandins, thromboxanes, and leukotrienes are mediators. The leukotrienes are formed by transformation of arachidonic acid into an unstable epoxide intermediate, leukotriene A4, which can
Evan S Dellon et al.
The American journal of gastroenterology, 107(10), 1503-1511 (2012-07-11)
Features of eosinophilic esophagitis (EoE) and gastroesophageal reflux disease (GERD) overlap. We aimed to determine whether staining for tissue biomarkers would differentiate EoE from GERD, suggesting utility for diagnosis of EoE. In this case-control study, EoE patients defined by consensus
J Maclouf et al.
Haemostasis, 26 Suppl 4, 28-36 (1996-10-01)
Formation of eicosanoids is a special mode of cell communication whereby production of eicosanoids by mixed cell populations differs from that expected from each individual cell. Transcellular biosynthesis of leukotriene C4 occurs via transfer of the reactive intermediate leukotriene A4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service