L4787
Leptin Receptor/Fc Chimera human
≥97% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture
Synonym(s):
B219
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
51111800
recombinant
expressed in NSO cells
assay
≥97% (SDS-PAGE)
form
lyophilized powder
technique(s)
cell culture | mammalian: suitable
UniProt accession no.
storage temp.
−20°C
Gene Information
human ... LEPR(3953)
General description
The gene LEPR (leptin receptor) is mapped to human chromosome 1p31. The neuroendocrine hormone, leptin, works through leptin receptor.
Biochem/physiol Actions
Leptin receptor (LEPR) is linked with risk of coronary artery disease. Mutation in LEPR is associated with low BMI (body mass index), increased leptin levels and some cancers. In humans and mice, problems in expression of leptin or leptin receptor are associated with obesity.
Leptin receptor (OB-R) is a single membrane-spanning receptor most related to the gp-130 signal-transducing component of the IL-6 receptor, the G-CSF receptor and the LIF receptor. OB-R, also known as B219, is expressed in the yolk sac, early fetal liver, enriched hematopoitic stem cells and in a variety of lymphohematopoietic cell lines. OB-R is also expressed at high levels in adult reproductive organs. OB-RL action in hepatic cells may be equivalent to that of IL-6 receptor. Leptin does not play a specific role in muting insulin action on hepatoma cells and may not contribute to the diabetic symptoms associated with obesity.
Physical form
Lyophilized from 20 mM Tris containing 5 mg of bovine serum albumin.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Song-Gen Jin et al.
International journal of clinical and experimental pathology, 8(5), 5968-5973 (2015-07-21)
The purpose of this study was to investigate the impact of the interactions among CX3CL1 (rs170364 and rs614230), LEPR (rs6700896), and IL-6 (rs2066992) polymorphisms on the risk of coronary artery disease (CAD) in Chinese Han population. 120 CAD patients and
Efraín Chavarria-Avila et al.
BioMed research international, 2015, 539408-539408 (2015-06-13)
The polymorphisms in leptin (LEP G-2548A) and leptin-receptor (LEPR Gln223Arg) seem to influence obesity and lipid metabolism among others. The aim of this study was to investigate the effect of these polymorphisms on adiposity, leptin (sLeptin), and leptin-receptor (sLeptin-receptor) serum
Jayasree Sreenivasan et al.
Immunology, 146(1), 122-129 (2015-06-11)
Leptin is an adipokine that regulates metabolism and plays an important role as a neuroendocrine hormone. Leptin mediates these functions via the leptin receptor, and deficiency in either leptin or its receptor leads to obesity in humans and mice. Leptin
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service