Key Documents
L4521
Sigma-Aldrich
Tamper-evident boxes
Replacement locks
About This Item
UNSPSC Code:
41123300
packaging
pack of 100 ea (locks)
General description
For transporting forensic evidence and medical samples with "chain of custody" assurance. Boxes are white plastic, of one-piece construction. They are sealed with plastic ties and numbered locks which must be broken to open.
Individually numbered
Choose from one of the most recent versions:
Certificates of Analysis (COA)