Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L3891

Sigma-Aldrich

Lactic dehydrogenase from rabbit muscle

Prestained, Molecular weight marker for SDS-PAGE

Synonym(s):

Lactate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

powder

mol wt

unstained protein 36,500 Da

storage temp.

2-8°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service