Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L3776

Sigma-Aldrich

Leukotriene C3

~50 μg/mL in methanol: water (7:3) containing 17 mM acetate buffer adjusted to pH 5.4 with NH4OH, ≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H49N3O9S
CAS Number:
Molecular Weight:
627.79
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

assay

≥97% (HPLC)

concentration

~50 μg/mL in methanol: water (7:3) containing 17 mM acetate buffer adjusted to pH 5.4 with NH4OH

functional group

carboxylic acid

shipped in

dry ice

Storage temp.

−20°C

SMILES string

CCCCCCCC\C=C\C=C\C=C\C(SCC(NC(=O)CCC(N)C(O)=O)C(=O)NCC(O)=O)C(O)CCCC(O)=O

InChI

1S/C30H49N3O9S/c1-2-3-4-5-6-7-8-9-10-11-12-13-16-25(24(34)15-14-17-27(36)37)43-21-23(29(40)32-20-28(38)39)33-26(35)19-18-22(31)30(41)42/h9-13,16,22-25,34H,2-8,14-15,17-21,31H2,1H3,(H,32,40)(H,33,35)(H,36,37)(H,38,39)(H,41,42)/b10-9+,12-11+,16-13+

Inchi Key

AIJDQMYBRDJHHT-FNBQXVLZSA-N

Packaging

Packaged under argon.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S E Dahlén et al.
European journal of pharmacology, 86(2), 207-215 (1982-12-24)
Eight biosynthetically formed cysteine-containing leukotrienes dose dependently (0.01-100 nM) contracted parenchymal strips of guinea-pig lung. The response to the leukotrienes was slow in onset, remarkably long-lasting and often tachyphylactic upon repeated administration. All the leukotrienes (LTC3, 8,9-LTC3, LTC4, 11-trans-LTC4, LTC5
Metabolism of leukotriene C3.
S Hammarström
Advances in prostaglandin, thromboxane, and leukotriene research, 9, 83-101 (1982-01-01)
C C Reddy et al.
Archives of biochemistry and biophysics, 279(2), 211-217 (1990-06-01)
Evidence for the formation of a positional isomer of leukotriene (LT) C3 (8,9-LTC3) from dihomo-gamma-linolenic acid has been published (Hammarström, S. J. Biol. Chem. 256, 7712-7714, 1981). This report describes the conversion of dihomo-gamma-linolenic acid to a postulated intermediate in
Biosynthesis and metabolism of leukotrienes.
S Hammarström
Monographs in allergy, 18, 265-271 (1983-01-01)
Rapid in vivo metabolism of leukotriene C3 in the monkey Macaca irus.
S Hammarström et al.
Biochemical and biophysical research communications, 101(4), 1109-1115 (1981-08-31)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service