L3413
[D-His(Bzl)6]-LH-RH Fragment 4-9 ethylamide trifluoroacetate salt
>95% (HPLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C44H64N12O8
Molecular Weight:
889.05
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32
Quality Level
assay
>95% (HPLC)
storage temp.
−20°C
Gene Information
human ... GNRH1(2796) , GNRHR(2798)
mouse ... GNRH1(14714) , GNRHR(14715)
rat ... GNRH1(25194) , GNRHR(81668)
Amino Acid Sequence
Ser-Tyr-His(Bz)-Leu-Arg-Pro-NHEt
Biochem/physiol Actions
Ser-Tyr-His(Bz)-Leu-Arg-Pro-NHEt ([D-His(Bzl)6]-LH-RH Fragment 4-9) is a peptide from GnRH (gonadotropin-releasing hormone) agonist, histerelin. GnRH (Glp-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2), which is also referred as LHRH (luteinizing hormone-releasing hormone) or gonadorelin, is crucial for mammalian reproduction and is released from hypothalamic neurons. It is responsible for the secretion of gonadotropins, luteinizing hormone (LH) and follicle-stimulating hormone (FSH), from the pituitary glands. Mutations in GnRH are associated with normosmic congenital hypogonadotropic hypogonadism. Histerelin is used for the treatment of central precocious puberty in children.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Amino Acids, Peptide and Proteins
Royal Society of Chemistry, 38 (2013)
Luigi Maione et al.
PloS one, 8(7), e69616-e69616 (2013-08-13)
Normosmic congenital hypogonadotropic hypogonadism (nCHH) is a rare reproductive disease leading to lack of puberty and infertility. Loss-of-function mutations of GNRH1 gene are a very rare cause of autosomal recessive nCHH. R31C GNRH1 is the only missense mutation that affects
Indira Nederpelt et al.
British journal of pharmacology, 173(1), 128-141 (2015-09-24)
Drug-target residence time is an important, yet often overlooked, parameter in drug discovery. Multiple studies have proposed an increased residence time to be beneficial for improved drug efficacy and/or longer duration of action. Currently, there are many drugs on the
Brooke Tata et al.
PLoS biology, 12(9), e1001952-e1001952 (2014-09-24)
Characterization of the genetic defects causing gonadotropic deficiency has made a major contribution to elucidation of the fundamental role of Kisspeptins and Neurokinin B in puberty onset and reproduction. The absence of puberty may also reveal neurodevelopmental disorders caused by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service