L3044
Monoclonal Anti-LRRK2 antibody produced in mouse
~1.0 mg/mL, clone PROK57, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-AURA17, Anti-Dardarin, Anti-Leucine-rich repeat protein kinase 2, Anti-PARK8, Anti-RIPK7, Anti-ROCO2
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
PROK57, monoclonal
form
buffered aqueous solution
mol wt
antigen ~250 kDa
species reactivity
mouse, human
concentration
~1.0 mg/mL
technique(s)
immunocytochemistry: suitable
indirect ELISA: suitable
western blot: 1-2 μg/mL using mouse brain crude extract
isotype
IgM
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... LRRK2(120892)
mouse ... Lrrk2(66725)
General description
Application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
related product
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service