Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

L2920

Sigma-Aldrich

Anti-LPP antibody, Mouse monoclonal

clone LPP4, purified from hybridoma cell culture

Synonym(s):

Anti-LIM domain-containing preferred translocation partner in lipoma, Anti-Lipoma-preferred partner

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

LPP4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

western blot: 0.5-1.0 μg/mL using whole extract of human HeLa cells

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LPP(4026)

General description

Monoclonal Anti-LPP (mouse IgG2a isotype) is derived from the hybridoma LPP4 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. Lipoma-preferred partner (LPP) gene encodes for the LPP protein, prominently expressed in smooth muscle. This protein contains a proline-rich N-terminal region and three C-terminally located LIN-11, Isl-1, and MEC-3 (LIM) domains. LPP protein is a member of the zyxin family. LPP is localized at sites of cell adhesion such as focal adhesions and cell-cell contacts. The LPP gene is mapped on the human chromosome at 3q27.3-q28.

Specificity

Monoclonal Anti-LPP recognizes human LPP.

Application

Anti-LPP antibody, Mouse monoclonal may be used in immunoblotting.

Biochem/physiol Actions

Lipoma-preferred partner (LPP) protein shuttles between cell adhesion sites and the nucleus where it has transcriptional activation capacity. LPP interacts with α-actin and VASP at cell adhesions via its N-terminal. It also interacts with tumor suppressor protein scrib at cell-cell contacts. The LPP gene is being rearranged in a subgroup of lipomas, which are benign tumors of adipose tissue. The LPP gene acts as a translocation partner in acute monoblastic leukemia.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze at –20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marleen M R Petit et al.
BMC cell biology, 6(1), 1-1 (2005-01-15)
At sites of cell adhesion, proteins exist that not only perform structural tasks but also have a signaling function. Previously, we found that the Lipoma Preferred Partner (LPP) protein is localized at sites of cell adhesion such as focal adhesions
W Huang et al.
Balkan journal of medical genetics : BJMG, 22(1), 5-10 (2019-09-17)
Copy number variation (CNV) is a main cause of male infertility, yet its influence still remains elusive in that of females. To investigate the correlation between CNV and female infertility, we applied whole-genome CNV analyses by next generation Sequencing (NGS)
Marleen M R Petit et al.
The Journal of biological chemistry, 278(4), 2157-2168 (2002-11-21)
Targeting of proteins to a particular cellular compartment is a critical determinant for proper functioning. LPP (LIM-containing lipoma-preferred partner) is a LIM domain protein that is localized at sites of cell adhesion and transiently in the nucleus. In various benign
M M Petit et al.
Molecular biology of the cell, 11(1), 117-129 (2000-01-19)
The LPP gene is the preferred translocation partner of the HMGIC gene in a subclass of human benign mesenchymal tumors known as lipomas. Here we have characterized the LPP gene product that shares 41% of sequence identity with the focal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service