L2766
Lectin from Datura stramonium (jimson weed, thorn apple)
lyophilized powder
Synonym(s):
Datura stramonium lectin, DSL
Select a Size
Select a Size
About This Item
Recommended Products
form
lyophilized powder
Quality Level
potency
≤10 μg per mL agglutination activity
composition
Protein, ≥60% biuret
impurities
salt, essentially free
storage temp.
−20°C
General description
Application
Biochem/physiol Actions
Quality
Preparation Note
Analysis Note
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service