L2662
Lectin from Viscum album (European mistletoe)
lyophilized powder
Synonym(s):
Viscum album agglutinin, VAA
About This Item
Recommended Products
form
lyophilized powder
potency
<16 μg/mL agglutination activity
composition
Protein, ~50% Lowry
storage temp.
−20°C
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
signalword
Danger
Hazard Classifications
Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Resp. Sens. 1 - Skin Sens. 1
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service