Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L2633

Sigma-Aldrich

Lectin from Ricinus communis (castor bean) Toxin RCA60–Peroxidase labeled

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

form

lyophilized powder

composition

Protein, ~75% modified Warburg-Christian

storage temp.

−20°C

Biochem/physiol Actions

Ricinus communis agglutinin (RCA) occurs in two forms designated RCA60 and RCA120 according to their mol. wt. of approx. 60,000 and 120,000 respectively.1 Neither RCA60 nor RCA120 is blood group specific. RCA60, also referred to as RCAII,2 Ricin D3 or RCL III4 is extremely toxic, inhibits protein synthesis and has an affinity for N-acetyl-D-galactosamine. RCA120, also referred to as RCAI2 or RCL (I + II)4 is an agglutinin and has an affinity for terminal β-D-galactosyl residues.1
Conjugates are prepared from affinity purified toxin, RCA60 or agglutinin RCA120.

Packaging

Package size based on protein content

Unit Definition

One unit will form 1 mg purpurogallin in 20 sec from pyrogallol at pH 6.0 at 20°C.

Physical form

Lyophilized powder containing sodium citrate

Preparation Note

Prepared from peroxidase type VI using two step glutaraldehyde method of Avrameas, S. and Ternyck, T., Immunochemistry, 8, 1175 (1971).
Repurified after conjugation by affinity chromatography.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 7.2, of a 1 mg protein per mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human erythrocytes after 1 hour incubation at 25°C.

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service