Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L2411

Sigma-Aldrich

LY-367,265

Synonym(s):

1-[2-[4-(6-fluoro-1H-indol-3-yl)-3,6-dihydro-1(2H)-pyridinyl]ethyl]-5,6-dihydro-1H,4H-[1,2,5]thiadiazolo[4.3.2-ij]quinoline-2,2-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

SMILES string

Fc1ccc2c(c[nH]c2c1)C3=CCN(CCN4c5cccc6CCCN(c56)S4(=O)=O)CC3

InChI

1S/C24H25FN4O2S/c25-19-6-7-20-21(16-26-22(20)15-19)17-8-11-27(12-9-17)13-14-28-23-5-1-3-18-4-2-10-29(24(18)23)32(28,30)31/h1,3,5-8,15-16,26H,2,4,9-14H2

InChI key

BJIPVHLRWSDKOS-UHFFFAOYSA-N

Gene Information

human ... HTR2A(3356)

Biochem/physiol Actions

Antidepressant, inhibits the serotonin transporter; 5-HT2A serotonin receptor antagonist.

Features and Benefits

This compound was developed by Eli Lilly. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service