Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L2408

Sigma-Aldrich

Lipoamide Dehydrogenase from torula yeast

lyophilized powder, 40-100 units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352204
Product L2408 is not currently sold in your country. Contact Technical Service

form

lyophilized powder

Quality Level

specific activity

40-100 units/mg protein

composition

Protein, ~80% Warburg-Christian

storage temp.

−20°C

Packaging

Sold on the basis of lipoamide dehydrogenase units.

Physical form

Lyophilized powder containing EDTA, phosphate and citrate buffer salts

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service