Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L1169

Sigma-Aldrich

Anti-Leukemia Inhibitory Factor antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

CDF, D factor, DIA, Emfilermin, LIF, differentiation inhibitory activity

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

immunohistochemistry: 25 μg/mL
neutralization: suitable
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... LIF(3976)

Looking for similar products? Visit Product Comparison Guide

General description

Leukemia inhibitory factor (LIF) is a pleotropic glycoprotein that belongs to the IL6 family of cytokines. It is secreted by many cell types such as, fibroblasts, macrophages, T cells, monocytes, liver cells, osteoblasts and cells of endometrium. LIF interacts with a heterodimeric receptor composed of two subunits, gp130 and gp190. A variety of signal transduction pathways are triggered in response to LIF including ERK/MAPK, JAK1, PI3K, STAT3, Jun-cFos and NF-κB. The various functions of LIF include proliferation of adult human T cells, maintenance of pluripotency of embryonic stem cells and inhibition of leukemic ell differentiation. Reports also indicate the important role of LIF in lactation, tissue regeneration after brain and spinal cord injury, muscle regeneration and motor neuron functions.
Anti-Leukemia Inhibitory Factor recognizes human leukemia inhibitory factor. It shows less than 30% cross-reactivity with recombinant mouse LIF.

Immunogen

purified, E. coli-derived recombinant human leukemia inhibitory factor.

Application

Anti-Leukemia Inhibitory Factor may be used for immunoblotting at a working a concentration of 0.1-0.2 μg/ml. For immunohistochemistry, a concentration of 25 μg/ml may be used. The antibody is also suitable for neutralization reactions.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L C Hunt et al.
Experimental cell research, 316(6), 1002-1009 (2009-12-08)
Leukemia inhibitory factor (LIF) is an important regulator of skeletal muscle regeneration and has been suggested to be mitogenic for myogenic cells because it has been shown to increase the quantity of myoblast cells grown in culture over extended periods
Ekaterini A Kritikou et al.
Development (Cambridge, England), 130(15), 3459-3468 (2003-06-18)
STAT3 is the key mediator of apoptosis in mammary gland. We demonstrate here that LIF is the physiological activator of STAT3, because in involuting mammary glands of Lif(-/-) mice, pSTAT3 is absent and the STAT3 target, C/EBPdelta, is not upregulated.
Sylvian Bauer et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 26(46), 12089-12099 (2006-11-17)
Although neural stem cells (NSCs) persist in various areas of the adult brain, their contribution to brain repair after injury is very limited. Treatment with exogenous growth factors can mitigate this limitation, suggesting that the brain environment is normally deficient
S M Metcalfe
Genes and immunity, 12(3), 157-168 (2011-03-04)
At the heart of lineage commitment within the adaptive immune response is the intrinsic genetic plasticity of the naive peripheral T lymphocyte (T cell). Primary activation by presentation of cognate antigen is coupled to rapid T-cell cycling and progressive epigenetic
D J Hilton
Trends in biochemical sciences, 17(2), 72-76 (1992-02-01)
Leukaemia inhibitory factor (LIF) is one of a growing number of cytokines that cannot be readily categorized according to its functions. Rather, these pleiotropic hormones have diverse and often overlapping effects on a multitude of cell types: for example, LIF

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service