Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L1040

Sigma-Aldrich

Livin β/ML-IAP human

>95% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in E. coli

Quality Level

assay

>95% (SDS-PAGE)

form

buffered aqueous solution

mol wt

32 kDa by SDS-PAGE

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... BIRC7(79444)

Biochem/physiol Actions

Livin β/ML-IAP, a member of the inhibitor of apoptosis (IAP) protein family, inhibits the activation of caspase 9 in cell extracts incubated with cytochrome c and dATP.
Member of the inhibitor of apoptosis (IAP) protein family; inhibits the activation of caspase 9 in cell extracts incubated with cytochrome c and dATP.

Physical form

Solution in 25 mM Hepes, pH 7.5, 0.1 M sodium chloride, and 1 mM dithiothreitol.

Analysis Note

Biological activity is measured by its ability to inhibit DEVD-afc cleavage in cell extracts activated by cytochrome c and dATP.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service