Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L1010

Sigma-Aldrich

Lung acetone powder from rat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Product L1010 is not currently sold in your country. Contact Technical Service

biological source

rat

form

powder

storage temp.

−20°C

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E A Wallnöfer et al.
Histochemistry and cell biology, 155(1), 19-73 (2020-10-12)
This multidisciplinary study examined the pharmacokinetics of nanoparticles based on albumin-DTPA-gadolinium chelates, testing the hypothesis that these nanoparticles create a stronger vessel signal than conventional gadolinium-based contrast agents and exploring if they are safe for clinical use. Nanoparticles based on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service