Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L0638

Sigma-Aldrich

Lactate Oxidase from Pediococcus sp.

lyophilized powder, ≥20 units/mg solid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

≥20 units/mg solid

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Formerly E.C. number 1.1.3.2
Lactate oxidase is a globular[1] flavoprotein and utilizes flavin mononucleotide (FMN) as the cofactor.[2] The enzyme contains a major tyrosine in the substrate-binding pocket[3] and a flavin adenine dinucleotide (FAD) active site.[4] Lactate oxidase has several bacterial sources.[1]

Application

Lactate Oxidase from Pediococcus sp. has been:
  • used to functionalize the detection reservoirs for lactate analysis[5]
  • used in sequential enzymatic reactions to measure lactate production in the supernatants of microglial cultures[6]
  • immobilized onto a carbon electrode to generate a self-powered lactate sensor[4]

Biochem/physiol Actions

Lactate oxidase is used in immobilization or enzyme-based labels for designing biochemical analytical devices such as immunosensors.[5][7] This enzyme catalyzes the oxidation of lactate to produce pyruvate and H2O2.[7] Lactate oxidase is useful in the first-generation biosensor approach for the electrochemical detection of lactate.[4]

Unit Definition

One unit will oxidize 1.0 μmole of L-lactate to pyruvate and H2O2 per min at pH 6.5 at 37 °C.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacob M Goran et al.
Analytical chemistry, 83(21), 8123-8129 (2011-09-29)
Nitrogen-doped carbon nanotubes (N-CNTs) provide a simple, robust, and unique platform for biosensing. Their catalytic activity toward the oxygen reduction reaction (ORR) and subsequent hydrogen peroxide (H(2)O(2)) disproportionation creates a sensitive electrochemical response to enzymatically generated H(2)O(2) on the N-CNT
Marcelo Ricardo Romero et al.
Analytical chemistry, 82(13), 5568-5572 (2010-06-04)
An amperometric sensor for lactate quantification is presented. The developed biosensor requires only 0.2 U of lactate oxidase, which is immobilized in a mucin/albumin hydrogel matrix. By protecting the platinum surface with a Nafion membrane, typical interference related to negatively
Xiang Shen et al.
Organic letters, 14(7), 1744-1747 (2012-03-27)
By making use of the aggregation-induced emission feature of silole 1 and the cascade reactions among l-lactic acid (LA), lactate oxidase (LOD), and dodecanoic hydrazine (DH), a new fluorometric "turn-on" method is developed for the detection of LA.
Parvin Samadi Pakchin et al.
Biosensors & bioelectronics, 122, 68-74 (2018-09-23)
In this work, a novel simple and sensitive electrochemical immunosensor was developed based on lactate oxidase as a single electrochemical probe for the detection of carcinoma antigen 125 (CA125). Chitosan-gold nanoparticle/multiwall carbon nanotube/graphene oxide (CS-AuNP/MWCNT/GO) was used as the electrode
Kavita Rathee et al.
Biochemistry and biophysics reports, 5, 35-54 (2015-11-11)
Lactate detection plays a significant role in healthcare, food industries and is specially necessitated in conditions like hemorrhage, respiratory failure, hepatic disease, sepsis and tissue hypoxia. Conventional methods for lactate determination are not accurate and fast so this accelerated the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service