Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L0377

Sigma-Aldrich

L-Lactic Dehydrogenase from bovine heart

Type IX, Predominantly the LDH H4 isotype, ammonium sulfate suspension, ≥300 units/mg protein

Synonym(s):

(S)-Lactate: NAD+ oxidoreductase, L-LDH, LAD, LD

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

type

Type IX

form

ammonium sulfate suspension

specific activity

≥300 units/mg protein

UniProt accession no.

foreign activity

glutamic-pyruvic transaminase <0.02%
pyruvate kinase, myokinase, glutamic-oxalacetic transaminase and α-glycerophosphate dehydrogenase <0.01%

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Also catalyzes the oxidation of other L-2-hydroxymonocarboxylic acids.

Unit Definition

One unit will reduce 1.0 μmole of pyruvate to L-lactate per min at pH 7.5 at 37 °C.

Physical form

Crystalline suspension in 2.1 M (NH4)2SO4 solution, pH 6.0

Analysis Note

Protein determined by biuret

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deshdeepak Sahni et al.
Journal of neurosurgery. Pediatrics, 11(5), 575-583 (2013-03-12)
Graphene possesses unique electrical, physical, and chemical properties that may offer significant potential as a bioscaffold for neuronal regeneration after spinal cord injury. The purpose of this investigation was to establish the in vitro biocompatibility of pristine graphene for interface
D Steffens et al.
Journal of biomedical nanotechnology, 9(4), 710-718 (2013-04-30)
The association of stem cells (SCs) with biomaterials promises to be the protagonist for future regenerative medicine in the treatment of tissue and organ lesions. Stem cells were cultivated in scaffolds constructed by the electrospinning technique, using poly-D,L-lactic acid (PDLLA)
Minh Vu Chuong Nguyen et al.
Biochemical pharmacology, 85(11), 1644-1654 (2013-04-16)
NADPH oxidase Nox4 is expressed in a wide range of tissues and plays a role in cellular signaling by providing reactive oxygen species (ROS) as intracellular messengers. Nox4 oxidase activity is thought to be constitutive and regulated at the transcriptional
Wei Quan et al.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan, 33(1), 85-91 (2013-04-20)
To determine the cardioprotective effect of magnesium lithospermate B (MLB) on myocardial ischemia/reperfusion (MI/R) injury and to investigate the antioxidant potential in vivo and in vitro. MI/R injury was induced by the occlusion of left anterior descending coronary artery for
O V Basharina et al.
Radiatsionnaia biologiia, radioecologiia, 52(6), 602-607 (2013-03-23)
The influence of UV-light (240-390 nm) at the dozes of 151 and 755 J/m2 on the intensity of processes of the lipid peroxidation, activity of lactate dehydrogenase (LDH), succinate dehydrogenase (SDH), cytochrome c oxidase and the level of the energy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service