Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L031030

Sigma-Aldrich

Activator 42®

0.1 M, configured for ABI

Synonym(s):

5-[3,5-Bis(trifluoromethyl)phenyl]-1H-tetrazole Solution, 5-[3,5-Bis(trifluoromethyl)phenyl]-2H-tetrazole Solution, Activator 42® Solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.21

type

for DNA synthesis

product line

Proligo Reagents

storage condition

dry at room temperature

concentration

28-21 g/L (Activator 42 content, titration)

impurities

≤30 ppm water content (Karl Fischer)

color

clear

compatibility

configured for ABI

InChI

1S/C9H4F6N4/c10-8(11,12)5-1-4(7-16-18-19-17-7)2-6(3-5)9(13,14)15/h1-3H,(H,16,17,18,19)

InChI key

KKJFZIQEWZVVIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Acetonitrile / Activator 42 100 / 3 (v/w)

Other Notes

Activator solution, containing 5-(3,5-bis(trifluoromethyl)phenyl)-1H-tetrazole (Activator 42) in acetonitrile, are mixed with solutions of phosphoramidites during the extension step.
The activator reacts with the amidite group to form a highly reactive intermediate. The intermediate then forms an internucleotide bond with the detritylated 5′hydroxyl group of the growing oligonucleotide chain.

Legal Information

This product and it use is are covered under US Patent Number 7,897,758
Activator 42 is a registered trademark of Sigma-Aldrich Co. LLC

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

41.0 °F

flash_point_c

5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service