Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L0300011-HH

ETT Activator

0.3M

Synonym(s):

5-(Ethylthio)-1H-tetrazole, 5-(Ethylthio)tetrazole solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

type

for DNA synthesis

product line

Proligo Reagents

packaging

pkg of 4 × 2.5 L L0302511-04

concentration

39-43 g/L (titration)

impurities

≤30 ppm water content (Karl Fischer)

color

clear Appearance of Solution

SMILES string

CCSc1nnn[nH]1

InChI

1S/C3H6N4S/c1-2-8-3-4-6-7-5-3/h2H2,1H3,(H,4,5,6,7)

InChI key

GONFBOIJNUKKST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Acetonitrile / 5-(Ethylthio)-1H- tetrazole 100 / 4 v/w

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service