Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

K4767

Sigma-Aldrich

Anti-PRKD1 (376-390) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-PKC-MU, Anti-PKCM, Anti-PKD, Anti-PRKCM, Anti-protein kinase D1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~102 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKD1(5587)

General description

Protein kinase D1 (PRKD1) belongs to the protein kinase D family. It is a serine/threonine kinase which is expressed in mammary glands. The gene encoding this protein is localized on human chromosome 14q12.

Immunogen

synthetic peptide corresponding to amino acids 376-390 of human PRKD1.

Application

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

Protein kinase D1 (PRKD1) has a role in insulin secretion in the islets of Langerhans. The protein has a role in various cellular processes like DNA synthesis, cell cycle progression, initiation of the nuclear factor κ-B pathway, vesicular trafficking and apoptosis. It conserves epithelial phenotype by inhibiting epithelial-to-mesenchymal transition. PRKD1 acts as an effector of diacylglycerol and protein kinase C.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel miR-34a target, protein kinase D1, stimulates cancer stemness and drug resistance through GSK3/?-catenin signaling in breast cancer
Do Yeon
Oncotarget, 7(12) (2016)
14q12 microdeletions excluding FOXG1 give rise to a congenital variant Rett syndrome-like phenotype
Carolyn Ellaway
European Journal of Human Genetics, 21(5) (2013)
Pharmacologic reversion of epigenetic silencing of the PRKD1 promoter blocks breast tumor cell invasion and metastasis.
Borges S
Breast Cancer Research, 15(2) (2013)
Molecular cloning, epigenetic regulation, and functional characterization of Prkd1 gene promoter in dopaminergic cell culture models of Parkinson's disease.
Ay M
Journal of Neurochemistry, 135(2), 402-415 (2015)
The PRKD1 promoter is a target of the KRas-NF-?B pathway in pancreatic cancer
Heike Doppler
Scientific Reports, 6 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service