Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K3264

Sigma-Aldrich

Anti-Ku80 (XRCC5) (418-439) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ATP-dependent DNA Helicase II, 80kDa, Anti-CTCBF (CTC85), Anti-Ku Autoantigen, 80kDa, Anti-Lupus Ku Autoantigen Protein p86, Anti-X-ray Repair Complementing Defective Repair in Chinese Hamster Cells 5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~82.5 kDa by calculation

species reactivity

human

technique(s)

western blot: 1:500

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... XRCC5(7520)
mouse ... Xrcc5(22596)
rat ... Xrcc5(363247)

Immunogen

peptide corresponding to the 80 kDa Ku80 protein (amino acids 418-439). This sequence is 100% conserved between human, mouse, rat, and hamster.

Physical form

Solution of 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service