Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K3019

Sigma-Aldrich

PKAcα, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC48865

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

1626-2201 nmol/min·mg

mol wt

~69 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKACA(5566)

Biochem/physiol Actions

Protein kinase A (PKA) is a mediator of cAMP (cyclic adenosine monophosphate) and contains two regulatory and two catalytic subunits. Mutations in PRKACA (protein kinase cAMP-activated catalytic subunit α) are associated with Cushing′s syndrome.[1] PRKACA encodes two splice variants, Cα1 and Cα2. Cα1 is widely expressed and Cα2 is restricted to the sperm cells.[2]

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Taskén et al.
Genomics, 36(3), 535-538 (1996-09-15)
We have determined the chromosomal localization of the gene for the catalytic subunit C alpha of cAMP-dependent protein kinase (locus PRKACA) to human chromosome 19 using polymerase chain reaction (PCR) and Southern blot analysis of two different somatic cell hybrid
Davide Calebiro et al.
Nature communications, 5, 5680-5680 (2014-12-06)
We recently identified a high prevalence of mutations affecting the catalytic (Cα) subunit of protein kinase A (PKA) in cortisol-secreting adrenocortical adenomas. The two identified mutations (Leu206Arg and Leu199_Cys200insTrp) are associated with increased PKA catalytic activity, but the underlying mechanisms
Bjørn S Skålhegg et al.
Molecular endocrinology (Baltimore, Md.), 16(3), 630-639 (2002-03-05)
The intracellular second messenger cAMP affects cell physiology by directly interacting with effector molecules that include cyclic nucleotide-gated ion channels, cAMP-regulated G protein exchange factors, and cAMP-dependent protein kinases (PKA). Two catalytic subunits, Calpha and Cbeta, are expressed in the
Maike M Vetter et al.
BMC biochemistry, 12, 40-40 (2011-08-05)
The two variants of the α-form of the catalytic (C) subunit of protein kinase A (PKA), designated Cα1 and Cα2, are encoded by the PRKACA gene. Whereas Cα1 is ubiquitous, Cα2 expression is restricted to the sperm cell. Cα1 and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service