Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K2260

Millipore

Kligler iron agar

pH 7.4±0.2 (25 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

powder

pH

7.4±0.2 (25 °C)

Application

Recommended as a differential medium in the study of the gram-negative intestinal microorganisms.

Other Notes

Components (g/L)
Peptic Digest of Animal Tissue 15.00
Beef Extract 3.00
Yeast Extract 3.00
Proteose Peptone 5.00
Lactose 10.00
Dextrose 1.00
Ferrous Sulphate 0.20
Sodium Chloride 5.00
Sodium Thiosulphate 0.30
Phenol Red 0.024
Agar 15.00

replaced by

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Isolation of Typhoid Bacilli from Urine and Feces with the Description of a new Double Sugar Tube Medium.
F F Russell
The Journal of medical research, 25(1), 217-229 (1911-09-01)
Bailey, S.F., et al.
Journal of Bacteriology, 13, 182-182 (1927)
A SIMPLE MEDIUM FOR THE DIFFERENTIATION OF MEMBERS OF THE TYPHOID-PARATYPHOID GROUP.
I J Kligler
American journal of public health (New York, N.Y. : 1912), 7(12), 1042-1044 (1917-12-01)
Kligler J.J.
Advances in Experimental Medicine and Biology, 28-319 (1918)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service