Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K1507

Sigma-Aldrich

Kirromycin from Streptomyces collinus

Synonym(s):

Antibiotic Myc-8003, Mocimycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H60N2O12
CAS Number:
Molecular Weight:
796.94
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

biological source

Streptomyces collinus

Quality Level

form

powder

color

yellow

solubility

methanol: 1.90-2.10 mg/mL, clear, yellow

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

CCC(C(=O)NC\C=C\C=C(/C)C(OC)C(C)C1OC(\C=C\C=C\C=C(/C)C(=O)C2=C(O)C=CNC2=O)C(O)C1O)C3(O)OC(\C=C\C=C\C)C(C)(C)C(O)C3O

InChI

1S/C43H60N2O12/c1-9-11-13-21-31-42(6,7)38(50)39(51)43(54,57-31)28(10-2)40(52)44-23-17-16-19-26(4)36(55-8)27(5)37-35(49)34(48)30(56-37)20-15-12-14-18-25(3)33(47)32-29(46)22-24-45-41(32)53/h9,11-22,24,27-28,30-31,34-39,48-51,54H,10,23H2,1-8H3,(H,44,52)(H2,45,46,53)/b11-9+,14-12+,17-16+,20-15+,21-13+,25-18+,26-19+

InChI key

HMSYAPGFKGSXAJ-DSMBXYKSSA-N

General description

Chemical structure: polyene

Application

Kirromycin is a polyketide antibiotic which is produced by the actinomycetes Streptomyces collinus. Kirromycin has been used to stimulate GTPase action of EF-Tu in the absence of programmed ribosomes in Thermus thermophilus[1]. Kirromycin is used to study bacterial protein synthesis at the level of elongation factor EF-Tu-GDP release.

Biochem/physiol Actions

Binding of kirromycin to EF-Tu prevents EF-TuGDP from being ejected from the ribosome, which immobilizes the ribsome, as well as all of the following ribosomes on the mRNA[2].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariorosario Masullo et al.
Biochemistry, 41(49), 14482-14488 (2002-12-05)
Valine 114 in the D(109)AAILVVA sequence of elongation factor 1alpha from the archaeon Sulfolobus solfataricus (SsEF-1alpha) was substituted with an acidic (V114E), basic (V114K), or cavity-forming (V114A) residue, and the effects on the biochemical properties of the factor were investigated.
H Rutthard et al.
The Journal of biological chemistry, 276(22), 18728-18733 (2001-03-29)
The influence of divalent metal ions on the intrinsic and kirromycin-stimulated GTPase activity in the absence of programmed ribosomes and on nucleotide binding affinity of elongation factor Tu (EF-Tu) from Thermus thermophilus prepared as the nucleotide- and Mg(2+)-free protein has
Subir Biswas et al.
International journal for parasitology, 41(3-4), 417-427 (2010-12-18)
Protein translation in the plastid (apicoplast) of Plasmodium spp. is of immense interest as a target for potential anti-malarial drugs. However, the molecular data on apicoplast translation needed for optimisation and development of novel inhibitors is lacking. We report characterisation
C C Hall et al.
Antimicrobial agents and chemotherapy, 33(3), 322-325 (1989-03-01)
Six kirromycin analogs (elfamycins) were compared on the basis of their inhibition of Escherichia coli poly(U)-directed poly(Phe) synthesis and stimulation of elongation factor Tu (EF-Tu)-associated GTPase activity. The elfamycins tested were kirromycin, aurodox, efrotomycin, phenelfamycin A, unphenelfamycin, and L-681,217. The
Lian N Olsthoorn-Tieleman et al.
Journal of bacteriology, 184(15), 4211-4218 (2002-07-11)
Streptomyces ramocissimus, the producer of elongation factor Tu (EF-Tu)-targeted antibiotic kirromycin, contains three divergent tuf-like genes, with tuf1 encoding regular kirromycin-sensitive EF-Tu1; the functions of tuf2 and tuf3 are unknown. Analysis of the tuf gene organization in nine producers of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service