Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K1014

Sigma-Aldrich

Anti-Kinesin 5B antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

western blot: 0.5 μg/mL using mouse neuronal cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... KIF5B(3799)
mouse ... Kif5b(16573)

General description

The gene KIF5B (kinesin 5B) is mapped to human chromosome 10p11.22. It encodes a conventional kinesin that contains two heavy chains and two light chains forming a heterotetramer. It is abundantly expressed in adipocytes.

Immunogen

synthetic peptide corresponding to amino acid residues 376-396 from human kinesin 5B.

Application

Anti-Kinesin 5B antibody produced in rabbit has been used in immunoprecipitation.[1]

Biochem/physiol Actions

The gene KIF5B (kinesin 5B) encodes a kinesin that is involved in the transportation of GLUT4 (glucose transporter protein) to the plasma membrane in response to insulin, thereby regulating insulin signaling pathway. It participates in the translocation of certain apical proteins to the apical membrane.

Physical form

Solution in phosphate buffered saline containing 1 mg/mL BSA and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polarization-dependent selective transport to the apical membrane by KIF5B in MDCK cells.
Jaulin F
Developmental Cell, 13, 511-522 (2007)
A transforming KIF5B and RET gene fusion in lung adenocarcinoma revealed from whole-genome and transcriptome sequencing.
Ju YS
Genome Research, 22, 436-445 (2012)
Conventional kinesin KIF5B mediates insulin-stimulated GLUT4 movements on microtubules.
Semiz S
The Embo Journal, 22, 2387-2399 (2003)
Raed Hanania et al.
The Journal of biological chemistry, 287(11), 8468-8483 (2012-01-25)
As major effector cells of the innate immune response, macrophages must adeptly migrate from blood to infected tissues. Endothelial transmigration is accomplished by matrix metalloproteinase (MMP)-induced degradation of basement membrane and extracellular matrix components. The classical activation of macrophages with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service