The antibody is directed against an intracellular epitope of C-terminal part of KCNE3.
Immunogen
synthetic peptide (C)RSRKVDKRSDPYH, corresponding to amino acid residues 81-93 of human KCNE3. The epitope is identical in rat, mouse, dog, and pig.
Physical form
Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA and 025% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.