Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

J4330

Sigma-Aldrich

JTE-907

≥98% (HPLC)

Synonym(s):

N-(1,3-benzodioxol-5-ylmethyl)-1,2-dihydro-7-methoxy-2-oxo-8-(pentyloxy)-3-Quinolinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26N2O6
CAS Number:
Molecular Weight:
438.47
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

color

white to off-white

solubility

DMSO: ≥10 mg/mL

Storage temp.

2-8°C

InChI

1S/C24H26N2O6/c1-3-4-5-10-30-22-19(29-2)9-7-16-12-17(24(28)26-21(16)22)23(27)25-13-15-6-8-18-20(11-15)32-14-31-18/h6-9,11-12H,3-5,10,13-14H2,1-2H3,(H,25,27)(H,26,28)

Inchi Key

GRAJFFFXJYFVOC-UHFFFAOYSA-N

Biochem/physiol Actions

JTE-907 is a cannabinoid CB2 ligand (inverse agonist).
JTE-907 is a cannabinoid CB2 ligand (inverse agonist). CB2 cannabanoid receptors are expressed in periferal nerves and other non CNS tissues, but are not expressed in the brain. CB2 is believed to have a role in nociceptive pain, but additional functions pertaining to the immune system have yet to be determined. CB2 selective agonists and antagonists provide a necessary tool for functional analyses.

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to cannabinoid receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service