Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

J3830

Sigma-Aldrich

JNJ 10191584 maleate salt

≥98% (HPLC)

Synonym(s):

1-[(5-Chloro-1H-benzimidazol-2-yl)carbonyl]-4-methylpiperazine maleate, VUF 6002 maleate, VUF6002 maleate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15ClN4O ·C4H4O4
CAS Number:
Molecular Weight:
394.81
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

off-white to light tan

solubility

DMSO: ≥10 mg/mL

originator

Johnson & Johnson

storage temp.

room temp

SMILES string

OC(=O)\C=C/C(O)=O.CN1CCN(CC1)C(=O)c2nc3cc(Cl)ccc3[nH]2

InChI

1S/C13H15ClN4O.C4H4O4/c1-17-4-6-18(7-5-17)13(19)12-15-10-3-2-9(14)8-11(10)16-12;5-3(6)1-2-4(7)8/h2-3,8H,4-7H2,1H3,(H,15,16);1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

KOTJFAYEELTYCZ-BTJKTKAUSA-N

Biochem/physiol Actions

JNJ 10191584 maleate is a highly selective histamine H4 receptor silent antagonist.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Johnson & Johnson. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to histamine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service