J3020
Anti-JAB1 antibody produced in rabbit
affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-Jun activation domain-binding protein-1
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 38 kDa
species reactivity
human, mouse, rat
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 10-20 μg/mL using sections of human breast carcinoma
immunoprecipitation (IP): 4-6 μg using RIPA extract (0.5 mg) of cultured rat pheochromocytoma PC12 cells
microarray: suitable
western blot: 0.25-0.5 μg/mL using whole extract of cultured mouse NIH3T3 fibroblasts
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... COPS5(10987)
mouse ... Cops5(26754)
rat ... Cops5(312916)
General description
Specificity
Immunogen
Application
- western blotting
- immunofluorescence
- immunoprecipitation
- immunohistology
Biochem/physiol Actions
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service