Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

J104

Sigma-Aldrich

Anti-c-Jun antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

western blot (chemiluminescent): 10 μg/mL using c-Jun expressed in serum stimulated mouse fibroblasts (NIH3T3)

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Jun(16476)

Immunogen

synthetic peptide corresponding to amino acid residues 91-105 of mouse c-Jun.

Application

Anti-c-Jun antibody produced in rabbit has been used as a primary antibody in immunohistochemistry.

Biochem/physiol Actions

The protooncogene c-Jun encodes a member of the jun family that forms a component of the transcription factor AP-1 (Activator Protein-1). It dimerizes with a member of the Fos family via the leucine-zipper motif to form AP-1, a factor involved in binding to TPA (12-O-tetradecanoylphorbol 13-acetate) response element (TRE) of various genes that include human collagenase, metallothionein IIa, stromelysin, interleukin 2, SV40 and polyoma. The dimers formed are involved in the activation of AP-1 dependent genes. AP-1 regulates the transcription of mammalian DDI (DNA-damage-inducible) genes.

Physical form

Lyophilized, purified rabbit IgG, containing 0.05% sodium azide.

Reconstitution

Reconstitute with 100 μL deionized water.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R F Paletzki et al.
Neuroscience, 152(4), 1040-1053 (2008-03-22)
We have expressed A-FOS, an inhibitor of activator protein-1 (AP-1) DNA binding, in adult mouse striatal neurons. We observed normal behavior including locomotion and exploratory activities. Following a single injection of cocaine, locomotion increased similarly in both the A-FOS expressing
P Angel et al.
Cell, 55(5), 875-885 (1988-12-02)
Binding of the human transcription factor Jun/AP-1 to a conserved 8 bp nucleotide sequence (TRE) is responsible for increased transcription of different cellular genes in response to tumor promoters, such as TPA, and serum factors. Enhanced Jun/AP-1 activity in TPA-stimulated
D A Brenner et al.
Nature, 337(6208), 661-663 (1989-02-16)
Tumour necrosis factor-alpha (TNF-alpha) is secreted by macrophages in response to inflammation, infection and cancer. Sublethal doses of recombinant TNF-alpha to rats causes cachexia, anaemia and inflammation. TNF-alpha plays a major part in tissue inflammation and remodelling by stimulating production
K Ryder et al.
Proceedings of the National Academy of Sciences of the United States of America, 86(5), 1500-1503 (1989-03-01)
The protooncogene c-jun encodes a component of the transcription factor AP-1. Both murine c-jun and a related gene (jun-B) are rapidly activated in BALB/c3T3 cells by serum growth factors. We report here the cloning and analysis of a cDNA encoding
Y Devary et al.
Molecular and cellular biology, 11(5), 2804-2811 (1991-05-01)
Exposure of mammalian cells to DNA-damaging agents leads to activation of a genetic response known as the UV response. Because several previously identified UV-inducible genes contain AP-1 binding sites within their promoters, we investigated the induction of AP-1 activity by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service