Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

IP0020

Millipore

Anti-c-Myc Immunoprecipitation Kit

sufficient for 50 assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.56

antibody product type

primary antibodies

usage

sufficient for 50 assays

analyte chemical class(es)

proteins (c-Myc)

technique(s)

immunoprecipitation (IP): suitable

storage temp.

2-8°C

General description

Immunoprecipitation is a method by which a protein can be specifically purified from a complex mixture of proteins using a specific antibody and a matrix that binds the antibody. The matrix bound protein can then be separated from the mixture by centrifugation. IP followed by SDS-PAGE and immunoblotting are routinely used in a variety of applications to measure the molecular masses of protein antigens, study protein interactions, determine specific enzymatic activity, monitor protein post-translation modifications, and determine the presence and quantity of proteins. The IP technique also enables the detection of rare proteins, which otherwise would be difficult to detect, since immunoprecipitation can concentrate them to as much as 10,000-fold.

Application

Specially designed to allow maximal recovery of c-Myc-tagged proteins in the immunoprecipitates. The whole process is performed in mini-spin columns, instead of in microcentrifuge tubes, thus combining convenient washing of the antigen-antibody-bound beads with minimal variability between experiments.
Anti-c-Myc immunoprecipitation kit was used in the identification of epithelial stromal interaction 1 as a novel effector downstream of Krüppel-like factor 8 in breast cancer invasion and metastasis.[1] It was also used to study the CD81 interaction with the T cell receptor to suppress signaling.[2]

Features and Benefits

  • Spin columns ensures more consistent results
  • Anti-c-Myc Affinity Gel ensures high specific binding to c-Myc-tagged fusion protein
  • CelLytic M Cell Lysis Reagent for rapid and efficient mammalian cell lysis

Legal Information

CelLytic is a trademark of Sigma-Aldrich Co. LLC

Not finding the right product?  

Try our Product Selector Tool.

Kit Components Also Available Separately

Product No.
Description
SDS

  • A7470Anti-c-Myc Agarose Affinity Gel antibody produced in rabbit, affinity isolated antibody 1 mLSDS

  • C2978CelLytic M, Cell Lysis Reagent, Suitable for Mammalian cell lysis and protein solubilization. 50 mLSDS

pictograms

CorrosionEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Medicine in Texas: the historical literature.
C R Burns
Texas medicine, 82(1), 60-63 (1986-01-01)
Safak Isil Cevik et al.
PloS one, 7(11), e50396-e50396 (2012-12-12)
CD81 (TAPA-1) is a ubiquitously expressed tetraspanin protein identified as a component of the B lymphocyte receptor (BCR) and as a receptor for the Hepatitis C Virus. In an effort to identify trans-membrane proteins that interact with the T-cell antigen
Dhanya Ravindran et al.
PloS one, 12(3), e0173224-e0173224 (2017-03-11)
Chemokines are important in macrophage recruitment and the progression of atherosclerosis. The 'M3' chemokine binding protein inactivates key chemokines involved in atherosclerosis (e.g. CCL2, CCL5 and CX3CL1). We aimed to determine the effect of M3 on plaque development and composition.
Wentong Li et al.
Scientific reports, 7(1), 300-300 (2017-03-24)
Dentin sialoprotein (DSP) is a dentin extracellular matrix protein. It is involved in dental mesenchymal cell lineages and dentin formation through regulation of its target gene expression. DSP mutations cause dentin genetic diseases. However, mechanisms of DSP in controlling dental
Liang Zhang et al.
Virulence, 11(1), 489-501 (2020-05-19)
Classical swine fever virus (CSFV), a positive-sense RNA virus, hijacks cell host proteins for its own replication. Rab18, a small Rab GTPase, regulates intracellular membrane-trafficking events between various compartments in cells and is involved in the life cycle of multiple

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service