Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ILGA4G3BCD

Sigma-Aldrich

SILuCarb Glycan Standard 13C Tetraantennary A4G3bcd

13C labeled asialo, mono-galactosylated tetraantennary complex N-glycan, Powered by Asparia Glycomics

Synonym(s):

SILuCarb Glycan Standard 13C Tetraantennary A4G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7813C6H140N6O61
Molecular Weight:
2215.97
UNSPSC Code:
41105331

shipped in

ambient

Storage temp.

2-8°C

General description

SILuCarb Glycan Standards are patented synthetic 13C-labeled N-glycans for use as internal standards in absolute glycan quantification by mass spectrometry.

Application

For use as a quantitative standard in applications such as Matrix-assisted Laser Desorption Ionization-time-of-flight (MALDI-TOF), Electrospray Ionization (ESI) and Liquid Chromatography Mass Spectrometry (LC-MS).

Physical form

13C labeled N-glycan refernce standard. 20 microM quantified solution

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service