I9881
Anti-Human IgG (γ-chain specific) antibody produced in goat
fractionated antiserum, lyophilized powder
About This Item
biological source
goat
conjugate
unconjugated
antibody form
fractionated antiserum
antibody product type
secondary antibodies
clone
polyclonal
form
lyophilized powder
packaging
vial of 2 mL lyophilized antiserum
technique(s)
Ouchterlony double diffusion: 1:16
storage temp.
2-8°C
target post-translational modification
unmodified
Looking for similar products? Visit Product Comparison Guide
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service