Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I9767

Sigma-Aldrich

Anti-IKKβ antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IκB Kinase β, Anti-IKK2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 85 kDa

species reactivity

human

technique(s)

western blot: 1:500 using a whole extract of human epitheloid carcinoma HeLa cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHUK(1147)

General description

IKK β is a component of I-κ-B-kinase (IKK) core complex consisting of IKK α, IKK β and IKK γ and belongs to Ser/Thr protein kinase family. It has a pivotal role in NF-κ-B signaling pathway. Anti-IKK β antibody can be used in immunoblot (diluted 1:1000). Rabbit anti-IKK β antibody reacts specifically with human IKK β (85 kD).

Immunogen

Synthetic peptide located near the C-terminal region of human IKKβ (amino acids 720-735) conjugated to KLH.

Application

Anti-IKK β antibody can be used in western blotting (diluted 1:500) by using whole extract of human epitheloid carcinoma HeLa cell line.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A DiDonato et al.
Nature, 388(6642), 548-554 (1997-08-07)
Nuclear transcription factors of the NF-kappaB/Rel family are inhibited by IkappaB proteins, which inactivate NF-kappaB by trapping it in the cell cytoplasm. Phosphorylation of IkappaBs marks them out for destruction, thereby relieving their inhibitory effect on NF-kappaB. A cytokine-activated protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service