Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I9410

Sigma-Aldrich

IsoQuick Strips for Mouse Monoclonal Isotyping

IgA, IgM

Synonym(s):

Mouse Monoclonal Antibody Isotyping Reagents, IgA, IgM Isotyping for Mouse Monoclonals

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Determination of the subclass of a monoclonal antibody is helpful for characterization of the antibody, for choosing detection reagents, and for deciding on a purification scheme.
The IsoQuick strips, whether purchased in kits (ISOQ5 and ISOQ25) or separately (I9285, I9410, or I9535), are designed to detect and identify mouse isotypes in cell culture supernatant, ascites, or purified antibody. Optimum detection range is 0.1-4 μg/mL with a detection limit of 10 ng/mL. IsoQuick strips have been validated for use with mouse monoclonal antibodies and have not been validated for use with other host antibodies.

Features and Benefits

The strips come ready-to-use and provide results in 5 minutes or less. The IsoQuick strips can be used individually or in combination with the same prepared sample for customized results.

Other Notes

The IsoQuick IgA/IgM strips are designed to detect and identify mouse IgA and IgM isotype subclasses.

Legal Information

IsoQuick is a trademark of EnviroLogix Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service