I9266
Insulin Receptor from rat liver
buffered aqueous glycerol solution
About This Item
biological source
rat liver
Quality Level
form
buffered aqueous glycerol solution
packaging
vial of 250 units
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
rat ... Insr(24954)
Biochem/physiol Actions
Unit Definition
Physical form
Preparation Note
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service