Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I9021

Sigma-Aldrich

Isomaltitol

≥98% (HPLC)

Synonym(s):

6-O-α-D-Glucopyranosyl-D-glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24O11
CAS Number:
Molecular Weight:
344.31
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

assay

≥98% (HPLC)

form

powder

sweetness

~0.6 × sucrose

impurities

≤8% water

color

white

solubility

water: 50 mg/mL, colorless

storage temp.

−20°C

SMILES string

OC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO[C@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

InChI

1S/C12H24O11/c13-1-4(15)7(17)8(18)5(16)3-22-12-11(21)10(20)9(19)6(2-14)23-12/h4-21H,1-3H2/t4-,5+,6+,7+,8+,9+,10-,11+,12-/m0/s1

InChI key

SERLAGPUMNYUCK-YJOKQAJESA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Sugar alcohol sweet tastant detectable by humans.

Other Notes

To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuji Nakamura et al.
PloS one, 12(10), e0186640-e0186640 (2017-10-19)
Cyclic nigerosyl nigerose (CNN) is a cyclic tetrasaccharide that exhibits properties distinct from other conventional cyclodextrins. Herein, we demonstrate that treatment of B16 melanoma with CNN results in a dose-dependent decrease in melanin synthesis, even under conditions that stimulate melanin
Kenji Abe et al.
Applied microbiology and biotechnology, 101(5), 2057-2066 (2016-12-10)
Industrial glucose feedstock prepared by enzymatic digestion of starch typically contains significant amounts of disaccharides such as maltose and isomaltose and trisaccharides such as maltotriose and panose. Maltose and maltosaccharides can be utilized in Escherichia coli fermentation using industrial glucose

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service