Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I8159

Sigma-Aldrich

Monoclonal Anti-Interleukin-1α/Interleukin-1F1 antibody produced in rat

clone 40508, purified immunoglobulin, lyophilized powder

Synonym(s):

Anti-Il1a

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

40508, monoclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

capture ELISA: 2 μg/mL
neutralization: suitable

UniProt accession no.

Storage temp.

−20°C

Gene Information

mouse ... Il1a(16175)

Immunogen

purified, E. coli-derived recombinant mouse interleukin-1α/interleukin-1F1.

Biochem/physiol Actions

Interleukins are proinflammatory cytokines produced on cell injury or trauma. There are two closely related types of interleukins produced by the cell, IL-1α and IL-1β that are produced in response to bacterial infection or LPS. Both IL-1α and IL-1β exert their cellular effects by signaling through IL-1 receptor type 1. The effects of IL-1α are mediated by activation of NF-κB, JNK and MAPK pathways that activated the genes in response to infection and inflammation. Overproduction of IL-1α has been implicated in inflammatory disorders like rheumatoid arthritis and periodontitis.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H E Barksby et al.
Clinical and experimental immunology, 149(2), 217-225 (2007-06-26)
Understanding cytokine immunobiology is central to the development of rational therapies for destructive inflammatory diseases such as rheumatoid arthritis (RA) and periodontitis. The classical interleukin-1 (IL-1) family cytokines, IL-1alpha and IL-1beta, as well as IL-18, play key roles in inflammation.
Axel Weber et al.
Science signaling, 3(105), cm1-cm1 (2010-01-21)
The interleukin-1 (IL-1) family of cytokines comprises 11 proteins (IL-1F1 to IL-1F11) encoded by 11 distinct genes in humans and mice. IL-1-type cytokines are major mediators of innate immune reactions, and blockade of the founding members IL-1alpha or IL-1beta by
Emmanuel Contassot et al.
Swiss medical weekly, 142, w13590-w13590 (2012-06-02)
Interleukin 1, one of the first cytokines discovered in the 1980s, and a potent mediator of fever, pain and inflammation, is at present experiencing a revival in biology and medicine. Whereas the mechanism of activation and secretion of interleukin 1β

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service